3-Methyl-1H-indazole-6-boronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-Methylindazole-6-boronic acid pinacol ester, 3-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

25692-1G 25692-25G 25692-500MG 25692-5G
76912-036EA 188.85 USD
76912-036 76912-038 76912-040 76912-042
3-Methyl-1H-indazole-6-boronic acid pinacol ester ≥95%
3-Methyl-1H-indazole-6-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BN₂O₂
MW: 258.1200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15071447
CAS Number: 1227911-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR