4-Bromo-2-chloro-5-fluoroaniline ≥97%

Supplier: AOBChem USA

Warning

25582-25G 25582-5G
76912-010EA 608.96 USD
76912-010 76912-012
4-Bromo-2-chloro-5-fluoroaniline ≥97%
4-Bromo-2-chloro-5-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrClFN
Storage Temperature: Ambient
MDL Number: MFCD09878155
CAS Number: 1000572-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR