3-Bromo-4-methylbiphenyl ≥97%

Supplier: AOBChem USA

Warning

25507-1G 25507-25G 25507-500MG 25507-5G
76911-996EA 194.15 USD
76911-996 76911-998 76912-000 76912-002
3-Bromo-4-methylbiphenyl ≥97%
3-Bromo-4-methylbiphenyl
Formula: C₁₃H₁₁Br
Storage Temperature: Refrigerator
MDL Number: MFCD23713125
CAS Number: 855255-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR