4-Bromo-2-(dimethylaminomethyl)-1-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

25195-1G 25195-25G 25195-500MG 25195-5G
76911-946EA 126.25 USD
76911-946 76911-948 76911-950 76911-952
4-Bromo-2-(dimethylaminomethyl)-1-fluorobenzene ≥97%
4-Bromo-2-(dimethylaminomethyl)-1-fluorobenzene
Formula: C₉H₁₁BrFN
MW: 232.093 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13249500
CAS Number: 188723-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR