3-Bromo-4-iodoanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-1-iodo-4-methoxybenzene

25158-25G 25158-5G
76911-920EA 268.41 USD
76911-920 76911-922
3-Bromo-4-iodoanisole ≥97%
3-Bromo-4-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆BrIO
Storage Temperature: Ambient
MDL Number: MFCD12547848
CAS Number: 466639-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR