4-Amino-3-fluorobiphenyl ≥97%

Supplier: AOBChem USA

Warning

25073-1G 25073-25G 25073-500MG 25073-5G
76911-900EA 290.69 USD
76911-900 76911-902 76911-904 76911-906
4-Amino-3-fluorobiphenyl ≥97%
4-Amino-3-fluorobiphenyl
Formula: C₁₂H₁₀FN
MW: 187.213 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06802824
CAS Number: 76302-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR