4-(6-Fluoro-3-pyridinyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

24907-1G 24907-500MG 24907-5G
76911-866EA 528.33 USD
76911-866 76911-868 76911-870
4-(6-Fluoro-3-pyridinyl)benzoic acid ≥97%
4-(6-Fluoro-3-pyridinyl)benzoic acid
Formula: C₁₂H₈FNO₂
MW: 217.196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11849236
CAS Number: 928658-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR