3-Fluoro-5-cyclopropylpyridine ≥97%

Supplier: AOBChem USA

Warning

24883-1G 24883-25G 24883-500MG 24883-5G
76911-850EA 225.98 USD
76911-850 76911-852 76911-854 76911-856
3-Fluoro-5-cyclopropylpyridine ≥97%
3-Fluoro-5-cyclopropylpyridine
Formula: C₈H₈FN
MW: 137.154 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22628235
CAS Number: 1414870-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR