3-Fluoro-5-methoxybenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,3,2-Dioxaborolane, 2-(3-fluoro-5-methoxyphenyl)-4,4,5,5-tetramethyl-, 3-Fluoro-5-methoxyphenylboronic acid pinacol ester, 2-(3-Fluoro-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

24844-1G 24844-25G 24844-5G
76911-826EA 62.6 USD
76911-826 76911-828 76911-830
3-Fluoro-5-methoxybenzeneboronic acid pinacol ester ≥97%
3-Fluoro-5-methoxybenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₃
Storage Temperature: Ambient
MDL Number: MFCD16996298
CAS Number: 1416367-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR