3-Chloro-2,4-difluorobenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

24780-25G 24780-5G
76911-812EA 453.01 USD
76911-812 76911-814
3-Chloro-2,4-difluorobenzyl alcohol ≥95%
3-Chloro-2,4-difluorobenzyl alcohol
Formula: C₇H₅ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD04116002
CAS Number: 886501-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR