3-Ethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

24719-1G 24719-25G 24719-5G
76911-788EA 62.6 USD
76911-788 76911-790 76911-792
3-Ethylbenzoic acid ≥97%
3-Ethylbenzoic acid
Formula: C₉H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00045851
CAS Number: 619-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR