3-Chloro-2,4-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24715-25G 24715-5G
76911-780EA 310.85 USD
76911-780 76911-782
3-Chloro-2,4-difluorobenzaldehyde ≥97%
3-Chloro-2,4-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD04116001
CAS Number: 127675-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR