4-Bromo-2-Chloro-5-(trifluoromethyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

24697-1G 24697-25G 24697-500MG 24697-5G
76911-770EA 260.99 USD
76911-770 76911-772 76911-774 76911-776
4-Bromo-2-Chloro-5-(trifluoromethyl)phenylboronic acid ≥95%
4-Bromo-2-Chloro-5-(trifluoromethyl)phenylboronic acid
Formula: C₇H₄BBrClF₃O₂
MW: 303.269 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628205
CAS Number: 1451393-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR