2-Acetamido-5-methoxytoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: N-(4-Methoxy-2-methylphenyl)acetamide, 4'-Methoxy-2'-methylacetanilide, 4-Acetamido-3-methylanisole

24681-25G 24681-5G
76911-758EA 325.7 USD
76911-758 76911-760
2-Acetamido-5-methoxytoluene ≥97%
2-Acetamido-5-methoxytoluene

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00458126
CAS Number: 31601-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR