3,5-Dimethylthioanisole ≥97%

Supplier: AOBChem USA

Warning

24644-1G 24644-25G 24644-5G
76911-742EA 78.51 USD
76911-742 76911-744 76911-746
3,5-Dimethylthioanisole ≥97%
3,5-Dimethylthioanisole

Precaución: for Research Use Only

Formula: C₉H₁₂S
Storage Temperature: Refrigerator
MDL Number: MFCD03789167
CAS Number: 66794-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR