3-Methoxy-5-methylpyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
24615-1G
24615-25G
24615-500MG
24615-5G
76911-732EA
290.69
USD
76911-732
76911-734
76911-736
76911-738
3-Methoxy-5-methylpyridine ≥97%
3-Methoxy-5-methylpyridine
Formula:
C₇H₉NO MW: 123.152 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11870766 CAS Number: 78210-42-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...