3-Bromo-5-(methylthio)benzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

24579-1G 24579-25G 24579-500MG 24579-5G
76911-702EA 201.58 USD
76911-702 76911-704 76911-706 76911-708
3-Bromo-5-(methylthio)benzoic acid ≥97% research grade
3-Bromo-5-(methylthio)benzoic acid
Formula: C₈H₇BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD22489506
CAS Number: 453566-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR