3-Chloro-5-(methylthio)benzoic acid ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
24564-1G
24564-25G
24564-500MG
24564-5G
76911-690EA
142.17
USD
76911-690
76911-692
76911-694
76911-696
3-Chloro-5-(methylthio)benzoic acid ≥97% research grade
3-Chloro-5-(methylthio)benzoic acid
Formula:
C₈H₇ClO₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD22489505 CAS Number: 53985-46-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...