3-Fluoro-5-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

24560-1G 24560-25G 24560-500MG 24560-5G
76911-678EA 257.8 USD
76911-678 76911-680 76911-682 76911-684
3-Fluoro-5-(methylthio)benzoic acid ≥97%
3-Fluoro-5-(methylthio)benzoic acid
Formula: C₈H₇FO₂S
Storage Temperature: Ambient
MDL Number: MFCD11976044
CAS Number: 453565-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR