4-(N,O-Dimethylhydroxylaminocarbonyl)-2-chlorophenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

24544-1G 24544-25G 24544-500MG 24544-5G
76911-664EA 379.81 USD
76911-664 76911-666 76911-668 76911-670
4-(N,O-Dimethylhydroxylaminocarbonyl)-2-chlorophenylboronic acid ≥97% research grade
4-(N,O-Dimethylhydroxylaminocarbonyl)-2-chlorophenylboronic acid
Formula: C₉H₁₁BClNO₄
Storage Temperature: Ambient
MDL Number: MFCD22543722
CAS Number: 1451392-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR