4-(5-Bromo-2-methoxybenzyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

24492-1G 24492-25G 24492-500MG 24492-5G
76911-626EA 320.4 USD
76911-626 76911-628 76911-630 76911-632
4-(5-Bromo-2-methoxybenzyl)morpholine ≥97%
4-(5-Bromo-2-methoxybenzyl)morpholine
Formula: C₁₂H₁₆BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD01307728
CAS Number: 7016-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR