3-Bromo-5-methylpyridine-2-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

24453-1G 24453-500MG 24453-5G
76911-612EA 706.56 USD
76911-612 76911-614 76911-616
3-Bromo-5-methylpyridine-2-carbaldehyde ≥95%
3-Bromo-5-methylpyridine-2-carbaldehyde
Formula: C₇H₆BrNO
MW: 200.033 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18254323
CAS Number: 1211520-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR