4,6-Dibromo-2-chloroanisole ≥97%

Supplier: AOBChem USA

Warning

24419-1G 24419-25G 24419-500MG 24419-5G
76911-596EA 292.81 USD
76911-596 76911-598 76911-600 76911-602
4,6-Dibromo-2-chloroanisole ≥97%
4,6-Dibromo-2-chloroanisole

Precaución: for Research Use Only

Formula: C₇H₅Br₂ClO
MW: 300.375 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22543700
CAS Number: 174913-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR