4-Amino-3,5-dichlorophenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

24393-1G 24393-25G 24393-500MG 24393-5G
76911-582EA 201.58 USD
76911-582 76911-584 76911-586 76911-588
4-Amino-3,5-dichlorophenylboronic acid, pinacol ester ≥97%
4-Amino-3,5-dichlorophenylboronic acid, pinacol ester
Formula: C₁₂H₁₆BCl₂NO₂
MW: 287.978 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23380454
CAS Number: 1371630-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR