4-[(2-Bromo-5-methoxyphenyl)methyl]morpholine ≥97%

Supplier: AOBChem USA

Warning

24377-1G 24377-25G 24377-500MG 24377-5G
76911-560EA 173.99 USD
76911-560 76911-562 76911-564 76911-566
4-[(2-Bromo-5-methoxyphenyl)methyl]morpholine ≥97%
4-[(2-Bromo-5-methoxyphenyl)methyl]morpholine
Formula: C₁₂H₁₆BrNO₂
MW: 286.165 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20994290
CAS Number: 1394291-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR