4-((5-Bromopyridin-2-yl)methyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

24312-1G 24312-25G 24312-500MG 24312-5G
76911-536EA 257.8 USD
76911-536 76911-538 76911-540 76911-542
4-((5-Bromopyridin-2-yl)methyl)morpholine ≥97%
4-((5-Bromopyridin-2-yl)methyl)morpholine
Formula: C₁₀H₁₃BrN₂O
MW: 257.127 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11977594
CAS Number: 294851-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR