3-Bromo-2,4-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24224-1G 24224-25G 24224-5G
76911-516EA 106.09 USD
76911-516 76911-518 76911-520
3-Bromo-2,4-difluorobenzaldehyde ≥97%
3-Bromo-2,4-difluorobenzaldehyde
Formula: C₇H₃BrF₂O
MW: 220.999 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22543686
CAS Number: 1326714-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR