4-(3,5-Dichlorophenyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24199-1G 24199-25G 24199-5G
76911-504EA 75.33 USD
76911-504 76911-506 76911-508
4-(3,5-Dichlorophenyl)benzaldehyde ≥97%
4-(3,5-Dichlorophenyl)benzaldehyde
Formula: C₁₃H₈Cl₂O
MW: 251.108 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01862518
CAS Number: 221018-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR