4-((6-Bromopyridin-3-yl)methyl)morpholine ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-[(6-Bromopyridine-3-yl)methyl]mopholine

23919-1G 23919-25G 23919-500MG 23919-5G
76911-414EA 252.5 USD
76911-414 76911-416 76911-418 76911-420
4-((6-Bromopyridin-3-yl)methyl)morpholine ≥97% research grade
4-((6-Bromopyridin-3-yl)methyl)morpholine
Formula: C₁₀H₁₃BrN₂O
MW: 257.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846170
CAS Number: 364793-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR