3,5-Dichloro-4-iodobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

23830-1G 23830-500MG 23830-5G
76911-390EA 530.45 USD
76911-390 76911-392 76911-394
3,5-Dichloro-4-iodobenzyl alcohol ≥97%
3,5-Dichloro-4-iodobenzyl alcohol
Formula: C₇H₅Cl₂IO
Storage Temperature: Ambient
CAS Number: 1350760-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR