3,4-Dibromo-2-methylaniline ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3,4-Dibromo-2-methyl-phenylamine

23710-1G 23710-25G 23710-500MG 23710-5G
76911-340EA 155.96 USD
76911-340 76911-342 76911-344 76911-346
3,4-Dibromo-2-methylaniline ≥97% research grade
3,4-Dibromo-2-methylaniline
Formula: C₇H₇Br₂N
Storage Temperature: Ambient
MDL Number: MFCD11845872
CAS Number: 858843-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR