3-Bromo-6-(4-trifluoromethylphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

23615-1G 23615-25G 23615-500MG 23615-5G
76911-320EA 421.18 USD
76911-320 76911-322 76911-324 76911-326
3-Bromo-6-(4-trifluoromethylphenyl)pyridine ≥97%
3-Bromo-6-(4-trifluoromethylphenyl)pyridine
Formula: C₁₂H₇BrF₃N
MW: 302.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14707263
CAS Number: 1215074-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR