3-Bromo-5-carboxythiophene-2-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

23539-1G 23539-25G 23539-500MG 23539-5G
76911-304EA 171.87 USD
76911-304 76911-306 76911-308 76911-310
3-Bromo-5-carboxythiophene-2-boronic acid ≥97% research grade
3-Bromo-5-carboxythiophene-2-boronic acid
Formula: C₅H₄BBrO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD22056771
CAS Number: 1451392-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR