3-Bromo-5-(ethoxymethyl)pyridine ≥95% research grade

Supplier: AOBChem USA

Warning

23496-1G 23496-25G 23496-500MG 23496-5G
76911-272EA 165.51 USD
76911-272 76911-274 76911-276 76911-278
3-Bromo-5-(ethoxymethyl)pyridine ≥95% research grade
3-Bromo-5-(ethoxymethyl)pyridine
Formula: C₈H₁₀BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD22056769
CAS Number: 723281-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR