4-(3-Bromobenzyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-[(3-Bromophenyl)methyl]morpholine

23438-1G 23438-25G 23438-500MG 23438-5G
76911-256EA 162.32 USD
76911-256 76911-258 76911-260 76911-262
4-(3-Bromobenzyl)morpholine ≥97%
4-(3-Bromobenzyl)morpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO
MW: 256.139 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00979456
CAS Number: 364793-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR