3-Iodo-2-phenoxypyridine ≥97%

Supplier: AOBChem USA

Warning

23397-1G 23397-25G 23397-500MG 23397-5G
76911-242EA 215.37 USD
76911-242 76911-244 76911-246 76911-248
3-Iodo-2-phenoxypyridine ≥97%
3-Iodo-2-phenoxypyridine
Formula: C₁₁H₈INO
MW: 297.092 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22045490
CAS Number: 754214-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR