3-(Piperidin-1-ylmethyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Piperidin-1-ylmethyl)phenylboronic acid, pinacol ester, 1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperidine

23329-1G 23329-25G 23329-5G
76911-204EA 65.78 USD
76911-204 76911-206 76911-208
3-(Piperidin-1-ylmethyl)phenylboronic acid pinacol ester ≥97%
3-(Piperidin-1-ylmethyl)phenylboronic acid pinacol ester
Formula: C₁₈H₂₈BNO₂
MW: 301.2314 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08435871
CAS Number: 859833-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR