4-Bromo-2-(trifluoromethoxy)phenol ≥97%

Supplier: AOBChem USA

Warning

23321-25G 23321-5G
76911-194EA 488.02 USD
76911-194 76911-196
4-Bromo-2-(trifluoromethoxy)phenol ≥97%
4-Bromo-2-(trifluoromethoxy)phenol

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09037798
CAS Number: 690264-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR