3-Fluoro-4-(methoxymethyl)phenylboronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

23185-1G 23185-500MG 23185-5G
76911-166EA 587.74 USD
76911-166 76911-168 76911-170
3-Fluoro-4-(methoxymethyl)phenylboronic acid ≥95% research grade
3-Fluoro-4-(methoxymethyl)phenylboronic acid
Formula: C₈H₁₀BFO₃
Storage Temperature: Ambient
MDL Number: MFCD14687259
CAS Number: 1313738-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR