3-Methyl-4-phenoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

23052-1G 23052-25G 23052-500MG 23052-5G
76911-130EA 173.99 USD
76911-130 76911-132 76911-134 76911-136
3-Methyl-4-phenoxybenzaldehyde ≥95%
3-Methyl-4-phenoxybenzaldehyde
Formula: C₁₄H₁₂O₂
MW: 212.244 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16717339
CAS Number: 191284-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR