3-Formyl-5-(trifluoromethyl)benzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

22971-1G 22971-25G 22971-500MG 22971-5G
76911-088EA 439.22 USD
76911-088 76911-090 76911-092 76911-094
3-Formyl-5-(trifluoromethyl)benzoic acid ≥97% research grade
3-Formyl-5-(trifluoromethyl)benzoic acid
Formula: C₉H₅F₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD22544381
CAS Number: 604001-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR