4-(4-Fluorobenzyl)morpholine ≥97%
Supplier: AOBChem USA
|
Warning
|
22851-1G
22851-25G
22851-500MG
22851-5G
76911-050EA
171.87
USD
76911-050
76911-052
76911-054
76911-056
4-(4-Fluorobenzyl)morpholine ≥97%
4-(4-Fluorobenzyl)morpholine
Formula:
C₁₁H₁₄FNO MW: 195.233 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00716890 CAS Number: 90754-65-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...