3,4-Difluoro-5-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

22842-1G 22842-25G 22842-500MG 22842-5G
76911-034EA 337.37 USD
76911-034 76911-036 76911-038 76911-040
3,4-Difluoro-5-formylphenylboronic acid ≥97%
3,4-Difluoro-5-formylphenylboronic acid
Formula: C₇H₅BF₂O₃
MW: 185.921 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22543655
CAS Number: 1451393-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR