3,5-Dichloro-4-iodotoluene ≥97%

Supplier: AOBChem USA

Warning

22816-25G 22816-5G
76911-022EA 453.01 USD
76911-022 76911-024
3,5-Dichloro-4-iodotoluene ≥97%
3,5-Dichloro-4-iodotoluene
Formula: C₇H₅Cl₂I
MW: 286.925 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526394
CAS Number: 939990-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR