3-Bromo-2-fluoro-6-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

22796-1G 22796-25G 22796-500MG 22796-5G
76911-002EA 296 USD
76911-002 76911-004 76911-006 76911-008
3-Bromo-2-fluoro-6-methylbenzaldehyde ≥97%
3-Bromo-2-fluoro-6-methylbenzaldehyde
Formula: C₈H₆BrFO
MW: 217.035 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26687661
CAS Number: 1518000-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR