3-Fluoro-5-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

22761-25G 22761-5G
76910-974EA 304.48 USD
76910-974 76910-976
3-Fluoro-5-methoxyphenol ≥97%
3-Fluoro-5-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇FO₂
MW: 142.1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848782
CAS Number: 850793-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR