3,5-Dichloro-4-ethoxyphenol ≥97%

Supplier: AOBChem USA

Warning

22759-1G 22759-25G 22759-500MG 22759-5G
76910-958EA 155.96 USD
76910-958 76910-960 76910-962 76910-964
3,5-Dichloro-4-ethoxyphenol ≥97%
3,5-Dichloro-4-ethoxyphenol
Formula: C₈H₈Cl₂O₂
MW: 207.054 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526388
CAS Number: 89748-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR