2,4-Dimethoxy-3-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

22757-1G 22757-500MG 22757-5G
76910-946EA 738.39 USD
76910-946 76910-948 76910-950
2,4-Dimethoxy-3-fluorobenzoic acid ≥97%
2,4-Dimethoxy-3-fluorobenzoic acid
Formula: C₉H₉FO₄
Storage Temperature: Ambient
MDL Number: MFCD24498657
CAS Number: 1018451-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR