3-Bromo-2-hydroxy-5-methoxybenzenecarbaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-2-hydroxy-5-methoxybenzaldehyde

22725-1G 22725-25G 22725-5G
76910-932EA 66.84 USD
76910-932 76910-934 76910-936
3-Bromo-2-hydroxy-5-methoxybenzenecarbaldehyde ≥97%
3-Bromo-2-hydroxy-5-methoxybenzenecarbaldehyde
Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00091938
CAS Number: 50343-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR