3-Methoxy-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

22535-25G 22535-5G
76910-864EA 318.27 USD
76910-864 76910-866
3-Methoxy-5-(trifluoromethyl)benzaldehyde ≥97%
3-Methoxy-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458144
CAS Number: 56723-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR